Learn More
Benzenesulfonic Acid Monohydrate, TRC
Specifications
Specifications
| Chemical Name or Material | Benzenesulfonic Acid Monohydrate |
| CAS | 26158-00-9 |
| Molecular Formula | C6H8O4S |
| Quantity | 50 g |
| Synonym | Benzenesulfonic Acid Hydrate |
| InChI Formula | InChI=1S/C6H6O3S.H2O/c7-10(8,9)6-4-2-1-3-5-6;/h1-5H,(H,7,8,9);1H2 |
| SMILES | O.OS(=O)(=O)c1ccccc1 |
| IUPAC Name | benzenesulfonic acid;hydrate |
| Molecular Weight (g/mol) | 176.19 |