Learn More
Benzo[c][1,2,5]oxadiazole, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 273-09-6 |
| Molecular Formula | C6H4N2O |
| MDL Number | MFCD00068064 |
| Quantity | 25 g |
| SMILES | O1N=C2C=CC=CC2=N1 |
| Molecular Weight (g/mol) | 120.11 |
| Percent Purity | 97% |
| Chemical Name or Material | Benzo[c][1,2,5]oxadiazole |