Learn More
Benzo[e]pyrene-d12, CDN
Specifications
Specifications
| Chemical Name or Material | Benzo[e]pyrene D12 |
| CAS | 205440-82-0 |
| Molecular Formula | C20 2H12 |
| Quantity | 0.01 g |
| Synonym | Benzo[e]pyrene-d12 (9 CI), Benzo(e)pyrene D12, Benzo[e]pyrene-1,2,3,4,5,6,7,8,9,10,11,12-d12 |
| InChI Formula | InChI=1 S/C20H12/c1-2-8-16-15(7-1)17-9-3-5-13-11-12-14-6-4-10-18(16)20(14)19(13)17/h1-12 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D,11 D,12 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c2c(c1[2 H])c3c([2 H])c([2 H])c([2 H])c4c([2 H])c([2 H])c5c([2 H])c([2 H])c([2 H])c2c5c34 |
| IUPAC Name | 1,2,3,4,5,6,7,8,9,10,11,12-dodecadeuteriobenzo[e]pyrene |
| Molecular Weight (g/mol) | 264.38 |
| Formula Weight | 264.1692 g/mol |
| Show More |