missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NucA nuclease Antibody, Clone: 1028905, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB100633-SP
This item is not returnable.
View return policy
Description
NucA Nuclease Monoclonal specifically detects NucA Nuclease in samples. It is validated for Western Blot, Simple Western.
Specifications
| NucA Nuclease | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 233874471 | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| 1028905 | |
| Western Blot 2 ug/mL, Simple Western 10 ug/mL | |
| P13717 | |
| E. coli-derived s. marcescens Benzonase Nuclease/NucA, Met1-Asn266, Accession # P13717 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction