missing translation for 'onlineSavingsMsg'
Learn More

Benzothiazole, TRC

Product Code. 18080830
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18080830 500 mg 500mg
18050430 1 g 1g
18062220 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18080830 Supplier Toronto Research Chemicals Supplier No. TRCB206640500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-16-9
Formula Weight 135.0143
InChI Formula InChI=1S/C7H5NS/c1-2-4-7-6(3-1)8-5-9-7/h1-5H
IUPAC Name 1,3-benzothiazole
Molecular Formula C7H5NS
Molecular Weight (g/mol) 135.19
Percent Purity >95%
SMILES c1ccc2scnc2c1
Synonym Benzothiazole, 1,3-Benzothiazole, 1-Thia-3-azaindene, Benzo[d]thiazole, Benzosulfonazole, NSC 8040, Vangard BT
Chemical Name or Material Benzothiazole
Grade HPLC
Show More Show Less

Specifications

Quantity 500 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.