missing translation for 'onlineSavingsMsg'
Learn More

Benzotrifluoride, 99%

Product Code. p-7025702
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
This item is not returnable. View return policy

Product Code. 11429267

Brand: Thermo Scientific Alfa Aesar B21340.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98-08-8 | C7H5F3 | 146.112 g/mol

It is applied as an alternative solvent to dichloromethane, where it has a higher bp. It is used as an intermediate for dyes and pharmaceuticals, as a solvent and a dielectric fluid, a vulcanizing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as an alternative solvent to dichloromethane, where it has a higher bp. It is used as an intermediate for dyes and pharmaceuticals, as a solvent and a dielectric fluid, a vulcanizing agent.

Solubility
Soluble in water at 20°C0.45g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-08-8
Molecular Formula C7H5F3
Molecular Weight (g/mol) 146.112
MDL Number MFCD00000372
InChI Key GETTZEONDQJALK-UHFFFAOYSA-N
Synonym benzotrifluoride, trifluoromethyl benzene, alpha,alpha,alpha-trifluorotoluene, phenylfluoroform, benzene, trifluoromethyl, benzenyl fluoride, benzylidyne fluoride, usaf ma-16, a,a,a-trifluorotoluene, trifluoro toluene
PubChem CID 7368
ChEBI CHEBI:36810
IUPAC Name trifluoromethylbenzene
SMILES C1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point -29°C
Density 1.188
Boiling Point 101°C to 103°C
Flash Point 12°C (54°F)
Odor Aromatic
Refractive Index 1.4145
Quantity 250 g
UN Number UN2338
Beilstein 1906908
Merck Index 14,1110
Solubility Information Soluble in water at 20°C0.45g/L.
Formula Weight 146.11
Percent Purity 99%
Chemical Name or Material Benzotrifluoride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Benzotrifluoride
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contains : Benzotrifluoride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.