missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl-2,3,4,5,6-d5-amine, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.10g
0.25g
Specifications
Specifications
| Chemical Name or Material | Benzylamine-2,3,4,5,6-D5 |
| CAS | 1219802-81-9 |
| Molecular Formula | C7 D5 H4 N |
| Quantity | 0.1 g |
| Synonym | Benzene-2,3,4,5,6-d5-methanamine (ACI), Benzyl-2,3,4,5,6-d5-amine, Benzylamine-d5, Benzylamine-2,3,4,5,6-D5, Benzylamine-d5 (phenyl-d5) |
| InChI Formula | InChI=1 S/C7H9N/c8-6-7-4-2-1-3-5-7/h1-5 H,6,8H2/i1D,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(CN)c([2 H])c1[2 H] |
| IUPAC Name | (2,3,4,5,6-pentadeuteriophenyl)methanamine |
| Molecular Weight (g/mol) | 112.1839 |
| Formula Weight | 112.1049 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction