missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl [(2-Benzylprop-2-enoyl)amino]acetate, Mikromol™
Description
Benzyl [(2-Benzylprop-2-enoyl)amino]acetate
Specifications
Specifications
| Chemical Name or Material | 2-Des(acetylthiomethyl)-2-methylene Racecadotril |
| CAS | 87428-99-7 |
| Analyte or Component Names | 2-Des(acetylthiomethyl)-2-methylene Racecadotril |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C19 H19 N O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 2-Des(acetylthiomethyl)-2-methylene Racecadotril, N-[1-Oxo-2-(phenylmethyl)-2-propen-1-yl]glycine Phenylmethyl Ester, N-[1-Oxo-2-(phenylmethyl)-2-propenyl]glycine Phenylmethyl Ester, Racecadotril Impurity F, Benzyl[(2-benzylprop-2-enoyl)amino]acetate, Racecadotril Impurity F |
| InChI Formula | InChI=1S/C19H19NO3/c1-15(12-16-8-4-2-5-9-16)19(22)20-13-18(21)23-14-17-10-6-3-7-11-17/h2-11H,1,12-14H2,(H,20,22) |
| SMILES | C=C(Cc1ccccc1)C(=O)NCC(=O)OCc2ccccc2 |
| IUPAC Name | benzyl 2-(2-benzylprop-2-enoylamino)acetate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?