missing translation for 'onlineSavingsMsg'
Learn More

Benzyl (5-Hydroxypentyl)carbamate, TRC

Product Code. 30311763
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30311763 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30311763 Supplier Toronto Research Chemicals Supplier No. TRCA5995805G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Amino-N-benzyloxycarbonylpentanol
CAS 87905-98-4
Molecular Formula C13 H19 N O3
Quantity 5 g
InChI Formula InChI=1S/C13H19NO3/c15-10-6-2-5-9-14-13(16)17-11-12-7-3-1-4-8-12/h1,3-4,7-8,15H,2,5-6,9-11H2,(H,14,16)
SMILES OCCCCCNC(=O)OCc1ccccc1
IUPAC Name benzyl N-(5-hydroxypentyl)carbamate
Molecular Weight (g/mol) 237.29
Formula Weight 237.1365
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.