Learn More
Berberine Chloride, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Berberine Chloride
Specifications
Specifications
| Chemical Name or Material | Berberine chloride |
| CAS | 633-65-8 |
| Analyte or Component Names | Berberine chloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H18 N O4 . Cl |
| Certifications/Compliance | ISO 9001 |
| Synonym | Benzo[g]-1,3-benzodioxolo[5,6-a]quinolizinium, 5,6-dihydro-9,10-dimethoxy-, chloride, Berberine, chloride, Berbinium, 7,8,13,13 A-tetradehydro-9,10-dimethoxy-2,3-(methylenedioxy)-, chloride, Berberine hydrochloride, Berberinium chloride, NSC 163088, NSC 646666 |
| InChI Formula | InChI=1 S/C20H18NO4.ClH/c1-22-17-4-3-12-7-16-14-9-19-18(24-11-25-19)8-13(14)5-6-21(16)10-15(12)20(17)23-2;/h3-4,7-10 H,5-6,11H2,1-2H3;1 H/q+1;/p-1 |
| SMILES | [Cl-].COc1ccc2cc3c4cc5OCOc5cc4CC[n+]3 cc2c1OC |
| IUPAC Name | 5,6-Dihydro-9,10-dimethoxybenzo[g]-1,3-benzodioxolo[5,6-a]quinolizinium chloride |
| Show More |