Learn More
Berkshire™ BCR™ Cleanroom Masks
Shop All Berkshire Products
Click to view available options
Unit Size:
Pack of 2500
Description
- Does not contain surfactants, fiberglass, glue, binders or thread
- Provide filtration efficiency and long wearing comfort for operators
- Soft, comfortable inside layer provides efficient particle and bacteria filtration and breathability
- Hems, seams, and attachments are ultrasonically bound for strength and cleanliness
- Cat. No 01-378 is individually wrapped and includes a clip to allow secure fastening behind head
- Cat. No 19-035-071 comes bulk packaged and is extra wide reducing the amount of exposed skin
Specifications
Specifications
| Product Type | Cleanroom Face Mask |
| Color | White |
| Fastening Type | Earloops |
| Material | Polypropylene |
| Style | Pleated |
| Cleanroom Class | Class 10 (ISO 4) |
| Construction | 3-ply |
| Model | BM.1FWE.50 |
| Series | BCR Series |