Learn More
Berkshire™ BCR™ Cleanroom Masks
Product Code. 16400134 Shop All Berkshire Products
Click to view available options
Unit Size:
Pack of 2500
Product Code. 16400134
Brand: Berkshire™ BM.1FWE.50
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed to protect cleanrooms and controlled environments from human contamination.
  • Does not contain surfactants, fiberglass, glue, binders or thread
  • Provide filtration efficiency and long wearing comfort for operators
  • Soft, comfortable inside layer provides efficient particle and bacteria filtration and breathability
  • Hems, seams, and attachments are ultrasonically bound for strength and cleanliness
  • Cat. No 01-378 is individually wrapped and includes a clip to allow secure fastening behind head
  • Cat. No 19-035-071 comes bulk packaged and is extra wide reducing the amount of exposed skin
Specifications
Product Type Cleanroom Face Mask
Color White
Fastening Type Earloops
Material Polypropylene
Style Pleated
Cleanroom Class Class 10 (ISO 4)
Construction 3-ply
Model BM.1FWE.50
Series BCR Series