Learn More
Berkshire™ SatPax™ 1000 Presaturated Wipers
Product Code. p-210196 Shop All Berkshire Products
Click to view available options
Container Type:
Resealable Pouch
Saturation Solution:
70% IPA, 30% Deionised Water
Packaging Quantity:
75/Pk.
Unit Size:
800g
Product Code. 12827153
Brand: Berkshire™ SPX1000.001.12
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For ISO Class 5 to 8 (100 to 100000) Cleanroom Class applications
  • Nonwoven hydroentangled 55% cellulose/45% polyester construction
  • Presaturated to eliminate the need to store, mix, and dispense solvents—no more squirt bottles or uncontrolled saturation levels
  • Manufactured using isopropyl alcohol and reverse osmosis/deionized water is filtered to 0.2μm — two saturation concentrations available
Order Info
Packaged in resealable pouches with lot numbers for traceability and canister dispensers with perforated rolls for popup convenience; canister refills available. Basis weight: 67.6g/cm2.
Specifications
Product Type IPA Wipe
Length (English) 9 in.
Length (Metric) 22.86 cm
Width (English) 9 in.
Width (Metric) 22.86 cm
Material Polyester Cellulose
Basis Weight 67.6
Color White
Container Type Resealable Pouch
Edges Knife-cut
Dimensions (L x W) 9 x 9 in. (22.86 x 22.86 cm)
For Use With (Application) For ISO Class 5 to 8 applications, presaturated to eliminate the need to store, mix, and dispense solvents no more squirt bottles or uncontrolled saturation levels
Packaging Quantity 75/Pk.
Saturation Solution 70% IPA, 30% Deionised Water
Sterility Non-sterile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Satpax 70% IPA/30% DI H2O
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H228-Flammable solid.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : propan-2-ol