missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BEST3 Monoclonal Antibody (OTI1H5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501878S
This item is not returnable.
View return policy
Description
BEST3 Monoclonal antibody specifically detects BEST3 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| BEST3 | |
| Monoclonal | |
| 0.75 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96C36 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 29920 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI1H5 | |
| Unconjugated | |
| Pycr2 | |
| BEST3; bestrophin 3; bestrophin-3; mBest4; vitelliform macular dystrophy 2-like 3; vitelliform macular dystrophy 2-like protein 3; Vmd2l3 | |
| Full length human recombinant protein of human PYCR2 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction