Learn More
Melford Bestatin HCl
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Competitive and specific inhibitor of leucine aminopeptidase, animo-peptidase B and triaminopeptidase.
Competitive and specific inhibitor of leucine aminopeptidase, animo-peptidase B and triaminopeptidase.
Specifications
Specifications
| Chemical Name or Material | Bestatin HCl |
| CAS | 65391-42-6 |
| Molecular Formula | C16H24N2O4 -HCl |
| Quantity | 10 mg |