missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Bestrophin 3 Antibody (OTI2H3), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72407C
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Bestrophin 3 Monoclonal specifically detects Bestrophin 3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Tekniske data
| Bestrophin 3 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| BEST3 | |
| Full length recombinant protein of human Bestrophin 3 (NP_689652) produced in HEK293T cells. | |
| 0.1 mL | |
| 144453.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI2H3 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| bestrophin 3, bestrophin-3, MGC13168, MGC40411, vitelliform macular dystrophy 2-like 3, VMD2L3Vitelliform macular dystrophy 2-like protein 3 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion