missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BET1 Polyclonal Antibody

Description
Immunogen sequence: MRRAGLGEGV PPGNYGNYGY ANSGYSACEE ENERLTESLR SKVTAIKSLS IEIGHEVKTQ NKLLAEMDSQ FDSTTGFLGK TMGKLKILSR GSQT; Positive Samples: U-87MG, HeLa; Cellular Location: Endoplasmic reticulum membrane, Golgi apparatus, Golgi apparatus membrane, Single-pass type IV membrane protein, cis-Golgi network membrane.
Specifications
Specifications
| Antigen | BET1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.66 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | BET1 |
| Gene Accession No. | O15155 |
| Gene Alias | BET1; Bet1 golgi vesicular membrane trafficking protein; BET1 homolog; Bet1p homolog; blocked early in transport 1 homolog; Golgi vesicular membrane trafficking protein p18; golgi vesicular membrane-trafficking protein p18; hBET1; rBET1 |
| Gene Symbols | BET1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?