missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-1,3-Glucuronyltransferase 1/B3GAT1 Antibody (3B8C12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 203.00 - € 552.00
Specifications
| Antigen | beta-1,3-Glucuronyltransferase 1/B3GAT1 |
|---|---|
| Clone | 3B8C12 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18239433
|
Novus Biologicals
NBP2-61709 |
0.1 mL |
€ 552.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18617216
|
Novus Biologicals
NBP2-61709-0.025ml |
0.025 mL |
€ 203.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
beta-1,3-Glucuronyltransferase 1/B3GAT1 Monoclonal specifically detects beta-1,3-Glucuronyltransferase 1/B3GAT1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
| beta-1,3-Glucuronyltransferase 1/B3GAT1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| 27087 | |
| Purified recombinant fragment of human B3GAT1 (AA: 193-334) expressed in E. Coli. | |
| Primary | |
| 38.2 kDa |
| 3B8C12 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 | |
| Monoclonal | |
| Purified | |
| Cancer, Immunology | |
| Beta-1,3-glucuronyltransferase 1, beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P), EC 2.4.1.135, galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1, GlcAT-P, GLCATPGLCUATP, glcUAT-P, Glucuronosyltransferase P, NK1, NK-1, UDP-GlcUA:glycoprotein beta-1,3-glucuronyltransferase | |
| B3GAT1 | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title