missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-1,3-Glucuronyltransferase 1/B3GAT1 Antibody (CL9499), Novus Biologicals™
Mouse Monoclonal Antibody
€ 401.00 - € 600.00
Specifications
| Antigen | beta-1,3-Glucuronyltransferase 1/B3GAT1 |
|---|---|
| Clone | CL9499 |
| Dilution | Western Blot 1 ug/ml, Immunocytochemistry/Immunofluorescence 2-10 ug/ml |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18694315
|
Novus Biologicals
NBP3-21170-100ul |
100 μg |
€ 600.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18611465
|
Novus Biologicals
NBP3-21170-25ul |
25 μg |
€ 401.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
beta-1,3-Glucuronyltransferase 1/B3GAT1 Monoclonal antibody specifically detects beta-1,3-Glucuronyltransferase 1/B3GAT1 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| beta-1,3-Glucuronyltransferase 1/B3GAT1 | |
| Western Blot 1 ug/ml, Immunocytochemistry/Immunofluorescence 2-10 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Beta-1,3-glucuronyltransferase 1, beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P), EC 2.4.1.135, galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1, GlcAT-P, GLCATPGLCUATP, glcUAT-P, Glucuronosyltransferase P, NK1, NK-1, UDP-GlcUA:glycoprotein beta-1,3-glucuronyltransferase | |
| This antibody was developed against a Synthetic Peptide of Q9P2W7. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| CL9499 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer, Immunology | |
| PBS, pH 7.2, 40% glycerol | |
| 27087 | |
| IgG2a | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title