Learn More
beta-1,3-Glucuronyltransferase 1/B3GAT1 Antibody (CL9500), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21171-25ul
 View more versions of this product
Product Code. 18606654
  • € 401.00 / 25µL
Description
beta-1,3-Glucuronyltransferase 1/B3GAT1 Monoclonal antibody specifically detects beta-1,3-Glucuronyltransferase 1/B3GAT1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
beta-1,3-Glucuronyltransferase 1/B3GAT1
Monoclonal
Unconjugated
PBS, pH 7.2, 40% glycerol
Mouse
Protein A purified
RUO
Primary
Human
Purified
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
CL9500
Western Blot 1 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Beta-1,3-glucuronyltransferase 1, beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P), EC 2.4.1.135, galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1, GlcAT-P, GLCATPGLCUATP, glcUAT-P, Glucuronosyltransferase P, NK1, NK-1, UDP-GlcUA:glycoprotein beta-1,3-glucuronyltransferase
This antibody was developed against a Synthetic Peptide of Q9P2W7. Actual immunogen sequence is proprietary.
25 μg
Cancer, Immunology
27087
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.