missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-1,3-Glucuronyltransferase 1/B3GAT1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-19788CL1
This item is not returnable.
View return policy
Description
beta-1,3-Glucuronyltransferase 1/B3GAT1 Polyclonal antibody specifically detects beta-1,3-Glucuronyltransferase 1/B3GAT1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| beta-1,3-Glucuronyltransferase 1/B3GAT1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| Beta-1,3-glucuronyltransferase 1, beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P), EC 2.4.1.135, galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1, GlcAT-P, GLCATPGLCUATP, glcUAT-P, Glucuronosyltransferase P, NK1, NK-1, UDP-GlcUA:glycoprotein beta-1,3-glucuronyltransferase | |
| A synthetic peptide made to an N-terminal portion of the human B3TGAT1 protein (between residues 1-50) [UniProt Q9P2W7] | |
| 0.1 mL | |
| Cancer, Immunology | |
| 27087 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction