missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta-1,3-N-Acetylglucosaminyltransferase 2/B3GNT2 Antibody (OTI3A9), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71624DL594
This item is not returnable.
View return policy
Description
Beta-1,3-N-Acetylglucosaminyltransferase 2/B3GNT2 Monoclonal antibody specifically detects Beta-1,3-N-Acetylglucosaminyltransferase 2/B3GNT2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, SDS-Page
Specifications
| Beta-1,3-N-Acetylglucosaminyltransferase 2/B3GNT2 | |
| Monoclonal | |
| DyLight 594 | |
| b3Gal-T7, B3GALT7, B3GNT, B3GNT1, B3GN-T1, B3GN-T2, B3GNT-2, Beta-1,3-galactosyltransferase 7, Beta-1,3-GalTase 7, beta-1,3-Gn-T1, Beta-1,3-Gn-T2, Beta-1,3-N-acetylglucosaminyltransferase 1, Beta-1,3-N-acetylglucosaminyltransferase 2, beta-1,3-N-acetylglucosaminyltransferase bGnT-1, beta-1,3-N-acetylglucosaminyltransferase bGnT-2, Beta3GalT7, Beta3Gal-T7, BETA3GNT, Beta3Gn-T1, beta3Gn-T2, Beta-3-Gx-T7, BGnT-1, BGnT-2, BGNT2, EC 2.4.1.-, UDP-Gal:beta-GlcNAc beta-1,3-galactosyltransferase 7, UDP-galactose:beta-N-acetylglucosamine beta-1,3-galactosyltransferase 7, UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 1, UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 2 | |
| Full length recombinant protein of human Beta-1,3-N-Acetylglucosaminyltransferase 2/B3GNT2 (NP_006568) produced in HEK293T cells. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 10678 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunofluorescence, SDS-Page | |
| OTI3A9 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction