missing translation for 'onlineSavingsMsg'
Learn More

beta-1,4-Galactosyltransferase 2/B4GalT2 Antibody, Novus Biologicals™

Product Code. p-200081186 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
25 μL
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18266523 100 μL 100µL
18637946 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18266523 Supplier Novus Biologicals Supplier No. NBP258906

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

beta-1,4-Galactosyltransferase 2/B4GalT2 Polyclonal specifically detects beta-1,4-Galactosyltransferase 2/B4GalT2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta-1,4-Galactosyltransferase 2/B4GalT2
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1-4 ug/ml
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias B4Gal-T2, B4Gal-T3, beta-1,4-galactosyltransferase 2, Beta-1,4-GalTase 2, beta-4-GalT2, beta4Gal-T2, beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase 2, EC 2.4.1.-, UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase 2, UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 2, UDP-Gal:betaGlcNAc beta 1,4-galactosyltransferase, polypeptide 2, UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 2, UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 2
Gene Symbols B4GALT2
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PYAGYFGGVSGLSKAQFLRINGFPNEYWGWGGEDDDIFNRISLTGMKISRPDIRIGRYRMIKHDRDKHNEPNPQRFTKIQNTKLTMKRDGIGSVRYQVLEVSRQPLFTNITV
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 8704
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.