missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Rat BNP-45 ELISA Kit (Colorimetric)

Product Code. 18612670 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
missing translation for 'unitSize'
1 set
This item is not returnable. View return policy

Product Code. 18612670

missing translation for 'mfr': Novus Biologicals™ NBP260599

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Kit (Colorimetric)

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 15.625 to 1000 pg/mL (example only, lot dependent)
Assay Sensitivity 0.025 ng/mL (example only; lot dependent)
Product Type ELISA Kit (Colorimetric)
Sample Type Serum, Plasma, Cell Culture, Tissue Extracts
Specificity This Rat BNP-45 ELISA Kit (Colorimetric) is specific to Brain Natriuretic Peptide 45 (aa 77-121).
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 4879
Gene Symbol NPPB
Quantity 1 Kit
Storage Requirements Storage is content dependent.
Synonym BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein
Target Rat
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.