missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta 2-Microglobulin Antibody (4G5A1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 218.00 - € 549.00
Specifications
| Antigen | beta 2-Microglobulin |
|---|---|
| Clone | 4G5A1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18639525
|
Novus Biologicals
NBP2-37284-0.025ml |
0.025 mL |
€ 218.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18791594
|
Novus Biologicals
NBP2-37284 |
0.1 mL | |||||||
Description
beta 2-Microglobulin Monoclonal specifically detects beta 2-Microglobulin in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| beta 2-Microglobulin | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin | |
| B2M | |
| IgG1 | |
| Affinity Purified |
| 4G5A1 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Adaptive Immunity, Cancer, Cell Biology, Diabetes Research, Immunology | |
| PBS with 0.05% Sodium Azide | |
| 567 | |
| Purified recombinant fragment of human B2M (AA: 21-100) expressed in E. Coli. | |
| Primary | |
| 13.7 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title