missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta 2-Microglobulin Antibody (883028) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8248JF585
This item is not returnable.
View return policy
Description
beta 2-Microglobulin Monoclonal antibody specifically detects beta 2-Microglobulin in Human samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| beta 2-Microglobulin | |
| Monoclonal | |
| Janelia Fluor 585 | |
| beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin | |
| Chinese hamster ovary cell line CHO-derived recombinant human β2-Microglobulin, Met1-Met119, Accession # P61769 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Western Blot, CyTOF | |
| 883028 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Adaptive Immunity, Cancer, Cell Biology, Diabetes Research, Immunology | |
| 567 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction