missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta 2-Microglobulin Antibody (B2M/961), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47704C
This item is not returnable.
View return policy
Description
beta 2-Microglobulin Monoclonal antibody specifically detects beta 2-Microglobulin in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Flow (Cell Surface), Immunofluorescence, CyTOF
Specifications
| beta 2-Microglobulin | |
| Monoclonal | |
| DyLight 650 | |
| beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin | |
| Full length recombinant human beta 2-Microglobulin protein (Uniprot: P61769 ) | |
| 0.1 mL | |
| Adaptive Immunity, Cancer, Cell Biology, Diabetes Research, Immunology | |
| 567 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| B2M/961 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction