missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD163 Antibody (EDHu-1), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-40686MFV500
This item is not returnable.
View return policy
Description
CD163 Monoclonal antibody specifically detects CD163 in Human, Rat, Porcine, Bovine, Guinea Pig, Monkey, Primate, Macaque, Sheep samples. It is validated for Flow Cytometry, Flow (Cell Surface)
Specifications
| CD163 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Rat, Pig, Bovine, Guinea Pig, Monkey, Primate | |
| Purified |
| Flow Cytometry | |
| EDHu-1 | |
| Flow Cytometry, Flow (Cell Surface) | |
| CD_antigen: CD163, CD163, CD163 molecule, Hemoglobin scavenger receptor, M130, macrophage-associated antigen, MM130, SCARI1, scavenger receptor cysteine-rich type 1 protein M130, sCD163, Soluble CD163 | |
| This CD163 Antibody (EDHu-1) was made from Leucocytes harvested from pleural cavity of patients with idiopathic spontaneous pneumothorax. | |
| 0.1 mL | |
| Angiogenesis, Immune System Diseases, Immunology, Inflammation, Innate Immunity | |
| 9332 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction