missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-3 Adrenergic R/ADRB3 Antibody (493324), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB9419CL1
This item is not returnable.
View return policy
Description
beta-3 Adrenergic R/ADRB3 Monoclonal antibody specifically detects beta-3 Adrenergic R/ADRB3 in Human samples. It is validated for Flow Cytometry,Immunocytochemistry
Specifications
| beta-3 Adrenergic R/ADRB3 | |
| Monoclonal | |
| CoraFluor 1 | |
| ADRB3R, adrenergic, beta-3-, receptor, B3AR, beta-3 adrenergic receptor, Beta-3 adrenoceptor, Beta-3 adrenoreceptor, BETA3AR, FLJ99960 | |
| Human embryonic kidney cell line HEK293-derived transfected with human beta-3 Adrenergic R/ADRB3, Met1-Ser408, Accession # P13945 | |
| 0.1 mL | |
| GPCR, Neuroscience | |
| 155 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Flow Cytometry, Immunocytochemistry | |
| 493324 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction