missing translation for 'onlineSavingsMsg'
Learn More

beta-Actin Antibody (ACTB/2370), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-20760AF700

Product Code. 18543438

  • € 728.00 / 0.10mL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

beta-Actin Monoclonal antibody specifically detects beta-Actin in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

beta-Actin
Monoclonal
Alexa Fluor 700
ACTB, actin, beta, actin, cytoplasmic 1, B-Actin, Beta Actin, beta cytoskeletal actin, Beta-actin, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1
Recombinant fragment (around aa150-350) of human beta-Actin protein (exact sequence is proprietary)
0.1 mL
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Loading Controls, Signal Transduction, Stem Cell Markers
60
Store at 4°C in the dark.
IgG
Immunohistochemistry (Paraffin)
ACTB/2370
50 mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.