missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Actin Antibody (ACTB/2370), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20760APC
This item is not returnable.
View return policy
Description
beta-Actin Monoclonal antibody specifically detects beta-Actin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| beta-Actin | |
| Monoclonal | |
| APC | |
| ACTB, actin, beta, actin, cytoplasmic 1, B-Actin, Beta Actin, beta cytoskeletal actin, Beta-actin, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1 | |
| Recombinant fragment (around aa150-350) of human beta-Actin protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Loading Controls, Signal Transduction, Stem Cell Markers | |
| 60 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| ACTB/2370 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction