Learn More
beta-Actin Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB600-503AF647
Product Code. 18726543
  • € 709.00 / 0.05mL
Description
beta-Actin Polyclonal specifically detects beta-Actin in Human, Mouse, Rat, Porcine, Avian, Bovine, Chinese Hamster, Fish, Primate, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
beta-Actin
Polyclonal
This beta-Actin Antibody was made from a synthetic peptide made to an N-terminal region of human Beta Actin. [UniProt P60709]
RUO
Store at 4C in the dark.
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry
Rabbit
0.05 mL
Pig