missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Amyloid Antibody (Kumamoto U. patent anti-Amyloid Beta), CoraFluor™ 1, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28292CL1
This item is not returnable.
View return policy
Description
beta Amyloid Monoclonal antibody specifically detects beta Amyloid in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| beta Amyloid | |
| Monoclonal | |
| CoraFluor 1 | |
| Abeta, Amyloid beta, beta-amyloid peptide | |
| Amyloid Beta | |
| 0.1 mL | |
| Alzheimers Research, Apoptosis, Cardiovascular Biology, Cell Cycle and Replication, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific, Signal Transduction | |
| 351 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Kumamoto U. patent anti-Amyloid Beta | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction