Learn More
Invitrogen™ beta Arrestin 1 Recombinant Rabbit Monoclonal Antibody (23GB1755), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA554204
 View more versions of this product
Product Code. 30089916
  • € 590.00 / 100µL
Description
Beta Arrestin 1 is a member of a family of proteins widely expressed but especially abundant in the central nervous system. Serving as an adaptor or scaffold molecule, beta Arrestin 1 is essential for mitogenic signaling and mediates agonist-dependent desensitization and internalization of Gprotein-coupled receptors (GPCRs, e.g., beta 2-adrenergic receptor). After binding to their ligand and interacting with heterotrimeric G proteins, GPCRs are phosphorylated by G-protein receptor kinases (GRKs) on serine residues. Beta Arrestin 1 in the cytosol is phosphorylated by ERK1 and 2 on serine412 in a negative feedback mechanism and binds to the phosphorylated receptors at the plasma membrane. Serine 412 is then dephosphorylated and the GPCRs are internalized, leading to activation of the Ras, Raf, ERK1 and 2 signaling pathway.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
beta Arrestin 1
Recombinant Monoclonal
Unconjugated
ARRB1
1200006I17Rik; ARB1; ARR1; ARRB1; ARRB1 protein; arrdc1; arrestin; arrestin 2; arrestin beta 1; arrestin beta-1; arrestin, beta 1; arrestin-2; AW208571; BARRES; beta-arrestin; beta-arrestin1; beta-arrestin-1; G430100A01Rik; Non-visual arrestin-2
Rabbit
Affinity chromatography
RUO
109689, 408
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
23GB1755
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P49407, Q8BWG8
ARRB1
A synthesized peptide derived from human ARRB1 (1-48AA).
100 μL
Primary
Human, Mouse