missing translation for 'onlineSavingsMsg'
Learn More

beta B1 Crystallin Antibody (2D8) [Alexa Fluor« 700], Novus Biologicals Biologicals™

Product Code. 30518953 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30518953 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30518953 Supplier Novus Biologicals Supplier No. NBP342949AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

beta B1 Crystallin Monoclonal antibody specifically detects beta B1 Crystallin in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta B1 Crystallin
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2D8
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias Beta-B1 crystallin, beta-crystallin B1, CATCN3, crystallin, beta B1, eye lens structural protein
Host Species Mouse
Immunogen crystallin, beta B1 - (Uniprot# P53674)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 1414
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.