missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Casein Antibody (F20.14), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2720F
This item is not returnable.
View return policy
Description
beta Casein Monoclonal specifically detects beta Casein in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| beta Casein | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| CSN2 | |
| Purified casein and a-lactalbumin. | |
| 0.1 mL | |
| Primary | |
| Does not cross react with alpha-lactalbumin. | |
| Store at 4C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Frozen) | |
| F20.14 | |
| Immunohistochemistry | |
| beta-casein, CASB, casein beta | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 1447 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction