Learn More
beta-Catenin Antibody (CTNNB1/6807R), Janelia Fluor™ 646, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-24344JF646
Code produit. 18504689
  • € 679.00 / 0.10mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
beta-Catenin Monoclonal antibody specifically detects beta-Catenin in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spécification
Spécification
beta-Catenin
Monoclonal
Janelia Fluor 646
beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein), beta 1, 88kDa, catenin beta-1, CTNNB, DKFZp686D02253, FLJ25606, FLJ37923
Recombinant fragment (around aa1-200) of human beta-Catenin protein (exact sequence is proprietary)
0.1 mL
Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Immune System Diseases, Mucosal Immunology, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway
1499
Store at 4°C in the dark.
IgG κ
Immunohistochemistry (Paraffin)
CTNNB1/6807R
50 mM Sodium Borate