missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Catenin Antibody (OTI12H7), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70509PE
This item is not returnable.
View return policy
Description
beta-Catenin Monoclonal specifically detects beta-Catenin in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Flow Cytometry.
Specifications
| beta-Catenin | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| CTNNB1 | |
| Full length recombinant protein of human beta-Catenin (NP_001895) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Primate, Monkey | |
| IgG1 |
| Flow Cytometry | |
| OTI12H7 | |
| Flow Cytometry | |
| beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein), beta 1, 88kDa, catenin beta-1, CTNNB, DKFZp686D02253, FLJ25606, FLJ37923 | |
| Mouse | |
| Affinity Purified | |
| Cancer, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Immune System Diseases, Mucosal Immunology, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
| 1499.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction