missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Catenin Antibody (rCTNNB1/8043), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24146G
This item is not returnable.
View return policy
Description
beta-Catenin Monoclonal antibody specifically detects beta-Catenin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| beta-Catenin | |
| Monoclonal | |
| DyLight 488 | |
| beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein), beta 1, 88kDa, catenin beta-1, CTNNB, DKFZp686D02253, FLJ25606, FLJ37923 | |
| Recombinant fragment (around aa1-200) of human beta-Catenin protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Immune System Diseases, Mucosal Immunology, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
| 1499 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rCTNNB1/8043 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction