Learn More
beta COP Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41358AF750
This item is not returnable.
View return policy
Description
beta COP Polyclonal antibody specifically detects beta COP in Human, Mouse samples. It is validated for Western Blot