missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta COP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41358
This item is not returnable.
View return policy
Description
beta COP Polyclonal antibody specifically detects beta COP in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| beta COP | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) | |
| beta coat protein, Beta-coat protein, beta-cop, coatomer protein complex, subunit beta, coatomer protein complex, subunit beta 1, coatomer subunit beta, COPBFLJ57957, DKFZp761K102, FLJ10341, FLJ46444 | |
| The epitope is recognized to a region between residue 903 to 853 of human Coatomer Protein Complex, Subunit Beta 1. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 μg/mg lysate | |
| NP_001137533.1 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 1315 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction