missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ beta COP Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA529655

Product Code. 13483867

  • € 522.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: Raji. Predicted reactivity: Mouse (100%), Rat (99%), Zebrafish (97%), Xenopus laevis (98%), Pig (100%), Chicken (99%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a protein subunit of the coatomer complex associated with non-clathrin coated vesicles. The coatomer complex, also known as the coat protein complex 1, forms in the cytoplasm and is recruited to the Golgi by activated guanosine triphosphatases. Once at the Golgi membrane, the coatomer complex may assist in the movement of protein and lipid components back to the endoplasmic reticulum. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

beta COP
Polyclonal
Unconjugated
Copb1
2610019B04Rik; beta coat protein; beta-coat protein; Beta-COP; coatomer protein complex subunit beta 1; coatomer protein complex, subunit beta 1; Coatomer subunit beta; COPB; COPB1; DKFZp761K102; FLJ10341; FLJ46444; FLJ57957; I79_013776; MSTP026; Rack2; receptor for activated C-kinase-2
Rabbit
Antigen affinity chromatography
RUO
114023, 1315
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.54 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P23514, P53618
Copb1
Recombinant protein encompassing a sequence within the N-terminus region of human COPB. The exact sequence is proprietary.
100 μL
Primary
Human, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.