Learn More
Beta Dystroglycan Antibody (P38G1) - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-43570
Product Code. 30279415
  • € 615.00 / 0.20mg
Description
Beta Dystroglycan Monoclonal antibody specifically detects Beta Dystroglycan in Human,Mouse samples. It is validated for Flow Cytometry,ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Beta Dystroglycan
Monoclonal
1 mg/mL
Flow Cytometry, ELISA
A3a, AGRNR, DAG156DAG, dystroglycan, dystroglycan 1 (dystrophin-associated glycoprotein 1), Dystrophin-associated glycoprotein 1
The original antibody was generated by immunizing BALB/c mice with a mixture of transfected HEK 293 cells expressing human and mouse beta-DAG. (Uniprot# Q14118; Q62165)
0.2 mg
Breast Cancer, Cancer
1605
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Flow Cytometry, ELISA
P38G1
Unconjugated
PBS
Rabbit
Protein A purified
RUO
Primary
Human, Mouse