missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Galactosidase Antibody (13R4), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52702AF647
This item is not returnable.
View return policy
Description
beta Galactosidase Monoclonal antibody specifically detects beta Galactosidase in E. coli samples. It is validated for Western Blot, Flow Cytometry, Control, CyTOF
Specifications
| beta Galactosidase | |
| Monoclonal | |
| Alexa Fluor 647 | |
| EC 3.2.1.23; b-gal | |
| Beta-galactosidase. | |
| 0.1 mL | |
| Epitope Tags | |
| E. coli | |
| Purified |
| Western Blot, Flow Cytometry, Control, CyTOF | |
| 13R4 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction