missing translation for 'onlineSavingsMsg'
Learn More

beta Galactosidase Antibody (13R4), CoraFluor™ 1, Novus Biologicals™

Human Monoclonal Antibody

Brand:  Novus Biologicals (Bio-Techne) NBP2-81032CL1

Product Code. 30142437

  • ONLINE EXCLUSIVE PRICE
    € 986.00
    € 932.40 / 0.10mL
    Save € 53.60
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

beta Galactosidase Monoclonal antibody specifically detects beta Galactosidase in E. coli samples. It is validated for Flow Cytometry,Western Blot,Control
TRUSTED_SUSTAINABILITY
Specifications

Specifications

beta Galactosidase
Monoclonal
CoraFluor 1
EC 3.2.1.23; b-gal
beta Galactosidase.
0.1 mL
Primary
Store at 4°C in the dark. Do not freeze.
IgG1 λ
Flow Cytometry, Western Blot, Control
13R4
PBS
Human
Protein A purified
RUO
E. coli
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.