missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Galactosidase Antibody, Biotin, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP1-76322B
This item is not returnable.
View return policy
Description
beta Galactosidase Polyclonal antibody specifically detects beta Galactosidase in E. coli samples. It is validated for Western Blot, ELISA
Specifications
| beta Galactosidase | |
| Polyclonal | |
| PBS | |
| Chicken | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgY |
| Western Blot, ELISA | |
| Biotin | |
| EC 3.2.1.23; b-gal | |
| Antibody was raised against a 17 amino acid synthetic peptide from near the amino terminus of E. coli beta-gal. The immunogen is located within amino acids 20 - 70 of beta-Galactosidase. Amino Acid Squence: WRNSEEARTDRPSQQLR | |
| 0.1 mL | |
| Epitope Tags, Fusion Proteins | |
| E. coli | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction