missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Galactosidase Antibody (DC1 4C7), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2619FR
This item is not returnable.
View return policy
Description
beta Galactosidase Monoclonal specifically detects beta Galactosidase in E. coli samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| beta Galactosidase | |
| Monoclonal | |
| DyLight 680 | |
| EC 3.2.1.23; b-gal | |
| Beta-galactosidase EC.3.2.1.23 from E.coli | |
| 0.1 mL | |
| Primary | |
| E. coli | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| DC1 4C7 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Beta-galactosidase (DC1 4C7). | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction