missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Galactosidase Antibody (DC1 4C7), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2619F
This item is not returnable.
View return policy
Description
beta Galactosidase Monoclonal specifically detects beta Galactosidase in E. coli samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| beta Galactosidase | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| Mouse | |
| Protein A purified | |
| RUO | |
| E. coli | |
| Purified |
| ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| DC1 4C7 | |
| ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| EC 3.2.1.23; b-gal | |
| Beta-galactosidase EC.3.2.1.23 from E.coli | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction