missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Glucuronidase/GUSB Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00002990-B01P
This item is not returnable.
View return policy
Description
beta-Glucuronidase/GUSB Polyclonal antibody specifically detects beta-Glucuronidase/GUSB in Human samples. It is validated for Western Blot
Specifications
| beta-Glucuronidase/GUSB | |
| Polyclonal | |
| Western Blot | |
| NP_000172.1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| beta-D-glucuronidase, beta-G1, beta-glucuronidase, BG, EC 3.2.1.31, FLJ39445, glucuronidase, beta, MPS7 | |
| GUSB (NP_000172.1, 1 a.a. - 651 a.a.) full-length human protein. MARGSAVAWAALGPLLWGCALGLQGGMLYPQESPSRECKELDGLWSFRADFSDNRRRGFEEQWYRRPLWESGPTVDMPVPSSFNDISQDWRLRHFVGWVWYEREVILPERWTQDLRTRVVLRIGSAHSYAIVWVNGVDTLEHEGGYLPFEADISNLVQVGPLPSRLRITIAINNTLTPTTLPPGTIQYLTDTSKYPKGYFVQNTYFDFFNYAGLQRSVLLYTTPTTYIDDITVTTSVEQDSGLVNYQISVKGSNLFKLEVRLLDAENKVVANGTGTQGQLKVPGVSLWWPYLMHERPAYLYSLEVQLTAQTSLGPVSDFYTLPVGIRTVAVTKSQFLINGKPFYFHGVNKHEDADIRGKGFDWPLLVKDFNLLRWLGANAFRTSHYPYAEEVMQMCDRYGIVVIDECPGVGLALPQFFNNVSLHHHMQVMEEVVRRDKNHPAVVMWSVANEPASHLESAGYYLKMVIAHTKSLDPSRPVTFVSNSNYAADKGAPYVDVICLNSYYSWYHDYGHLELIQLQLATQFENWYKKYQKPIIQSEYGAETIAGFHQDPPLMFTEEYQKSLLEQYHLGLDQKRRKYVVGELIWNFADFMTEQSPTRVLGNKKGIFTRQRQPKSAAFLLRERYWKIANETRYPHSVAKSQCLENSPFT | |
| 0.05 mg | |
| Cancer, Endocrinology, Signal Transduction | |
| 2990 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction