missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta II Tubulin Antibody (OTI3B7), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
€ 256.00 - € 550.00
Specifications
| Antigen | beta II Tubulin |
|---|---|
| Clone | OTI3B7 |
| Dilution | Western Blot 1:4000, Immunocytochemistry/ Immunofluorescence 1:100 |
| Applications | Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306496
|
Novus Biologicals
NBP3-44865-100ul |
100 μL |
€ 550.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306647
|
Novus Biologicals
NBP3-44865-30ul |
30 μL |
€ 256.00
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
beta II Tubulin Monoclonal antibody specifically detects beta II Tubulin in Human,Mouse,Rat samples. It is validated for Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
| beta II Tubulin | |
| Western Blot 1:4000, Immunocytochemistry/ Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| IgG2a | |
| >95% | |
| Antibody |
| OTI3B7 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| Full length human recombinant protein of human TUBB2A(NP_001060) produced in HEK293T cell. | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title