missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta II Tubulin B Antibody (5B2), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-74037H
This item is not returnable.
View return policy
Description
beta II Tubulin B Monoclonal specifically detects beta II Tubulin B in Human samples. It is validated for Western Blot, ELISA.
Specifications
| beta II Tubulin B | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| TUBB2B | |
| Recombinant human beta-Tubulin (1-445aa) purified from E. coli | |
| 100 μL | |
| Signal Transduction | |
| 347733 | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA | |
| 5B2 | |
| Western Blot, ELISA | |
| bA506K6.1, class II beta-tubulin isotype, DKFZp566F223, FLJ98847, MGC8685, tubulin beta-2B chain, tubulin, beta 2B, tubulin, beta polypeptide paralog | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human beta-Tubulin mAb, clone 5B2, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human beta-Tubulin protein. | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction